Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: 1810014B01Rik | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_015572.2 | 3 | 1051 | |||
GenBank mRNA | AK007490.1 | - | 3 | 1052 | ||
AK012122.1 | - | 3 | 1097 | |||
AK049515.1 | - | 2 | 1069 | |||
AK090016.1 | - | 3 | 1095 |