Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Prkab1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_031976.1 | NP_114182.1 | 4 | 570 | ||
XM_006249481.2 | XP_006249543.1 | 5 | 570 | |||
GenBank mRNA | BC062008.1 | - | 4 | 456 | 5599996 | |
U42411.1 | - | 4 | 570 | |||
X95577.1 | - | 5 | 540 |