Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ctnnbip1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001173388.1 | NP_001166859.1 | 4, 5 | 421 | ||
XM_006239436.2 | XP_006239498.1 | 3, 4 | 408 | |||
GenBank mRNA | BC098773.1 | - | 3, 4 | 271 | ||
CB611569.1 | - | 4, 5 | 438 |