Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Col13a1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001109172.1 | NP_001102642.1 | 16, 17 | 1368 | ||
XM_006256470.2 | XP_006256532.2 | 15, 16 | 1359 | |||
XM_006256479.2 | XP_006256541.1 | 15, 16 | 1367 | |||
XM_006256481.2 | XP_006256543.1 | 12, 13 | 1237 | |||
XM_008772904.1 | XP_008771126.1 | 15, 16 | 1368 | |||
XM_008772905.1 | XP_008771127.1 | 14, 15 | 1341 | |||
XM_008772906.1 | XP_008771128.1 | 15, 16 | 1368 | |||
XM_008772907.1 | XP_008771129.1 | 15, 16 | 1368 | |||
XM_008772908.1 | XP_008771130.1 | 14, 15 | 1332 | |||
XM_008772909.1 | XP_008771131.1 | 13, 14 | 1305 | |||
XM_008772910.1 | XP_008771132.1 | 14, 15 | 1302 | |||
XM_008772911.1 | XP_008771133.1 | 15, 16 | 1369 | |||
XM_008772912.1 | XP_008771134.1 | 12, 13 | 1254 | |||
GenBank mRNA | BC162018.1 | - | 12, 13 | 1002 | 9098304 |