Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: aicda | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001100779.1 | NP_001094249.1 | 4 | 579 | ||
GenBank mRNA | AJ606071.2 | - | 3 | 400 |