Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Bfar | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001013125.1 | NP_001013143.1 | 3 | 496 | ||
GenBank mRNA | BC087103.1 | - | 3 | 496 | 7192877 |