Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Enterprise Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search Thermo Fisher Scientific

Search All
Search button Close
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          See other MAPK8IP3 SIRNA Assays ›
          Gene Symbol:
          Mapk8ip3
          Gene Name:
          mitogen-activated protein kinase 8 interacting protein 3
          Gene Aliases:
          JIP3, JSAP1
          Chromosome Location:
          Chr.10: 14259394 - 14299276 on Build Rnor_6.0
          UniGene ID:
          Rn.16158
          Species:
          Rat
          siRNA ID RSS357211
          Size 20 nmol
          Availability Made To Order
          Catalog # 1330001
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • siRNA Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          siRNA Details

          Gene Symbol: Mapk8ip3
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
          RefSeq NM_001100673.1 NP_001094143.1 14, 15 1685
          XM_006245953.2 XP_006246015.1 16, 17 1863
          XM_006245954.2 XP_006246016.1 16, 17 1860
          XM_006245955.2 XP_006246017.1 15, 16 1838
          XM_006245956.2 XP_006246018.1 15, 16 1836
          XM_006245957.2 XP_006246019.1 16, 17 1836
          XM_006245958.2 XP_006246020.1 14, 15 1817
          XM_006245959.2 XP_006246021.1 16, 17 1770
          XM_006245960.2 XP_006246022.1 16, 17 1767
          XM_006245961.2 XP_006246023.1 15, 16 1745
          XM_006245962.2 XP_006246024.1 15, 16 1742
          XM_006245963.2 XP_006246025.1 14, 15 1727
          XM_008767567.1 XP_008765789.1 15, 16 1844
          XM_008767568.1 XP_008765790.1 11, 12 1535
          XM_008767569.1 XP_008765791.1 11, 12 1072
          GenBank mRNA DQ377222.1 - 14, 15 1685

          Back To Top

          More Information


          Gene Ontology Categories:

          Function(s) Process(es)

          in utero embryonic development
          JNK cascade
          activation of JUN kinase activity
          axon guidance
          respiratory gaseous exchange
          protein localization
          post-embryonic development
          positive regulation of signal transduction
          regulation of gene expression
          vesicle-mediated transport
          forebrain development
          neuron projection development
          positive regulation of JUN kinase activity
          positive regulation of neuron differentiation
          regulation of JNK cascade
          lung alveolus development
          lung morphogenesis
          MAP-kinase scaffold activity
          JUN kinase binding
          kinesin binding
          receptor signaling complex scaffold activity
          mitogen-activated protein kinase kinase binding
          mitogen-activated protein kinase kinase kinase binding

          Back To Top

          Related Products

          • Anti-GAPDH, Mouse Monoclonal 6C5
          • Anti-β-Actin, Mouse Monoclonal AC-15
          • TaqMan® Gene Expression Cells-to-CT™
          • Neon™ Transfection System 100 µL Kit
          • Lipofectamine™ RNAiMAX Transfection Reagent
          • Invivofectamine 2.0
          • Lipofectamine™ 2000 Transfection Reagent
          • Neon™ Transfection System
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us
          • Careers
          • Investors
          • News
          • Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2025 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-666774c847-nqx77:80/100.66.134.75:80.
          git-commit: 4f7bff5b1d64937db6e1726f71785770340a1908
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.34.0-2025.08.32-1.0