Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Gpx4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001039849.2 | NP_001034938.1 | 3, 4 | 505 | ||
NM_017165.2 | NP_058861.3 | 3, 4 | 391 | |||
GenBank mRNA | AB072798.1 | - | 3, 4 | 505 | ||
AF274028.1 | - | 3, 4 | 514 | |||
AY570513.1 | - | 3, 4 | 493 | |||
CB582508.1 | - | 3, 4 | 415 | |||
FQ210610.1 | - | 3, 4 | 299 | |||
FQ221663.1 | - | 3, 4 | 321 | |||
FQ221760.1 | - | 3, 4 | 299 | |||
FQ221899.1 | - | 3, 4 | 299 | |||
FQ221976.1 | - | 3, 4 | 302 | |||
FQ222299.1 | - | 3, 4 | 299 | |||
FQ227564.1 | - | 3, 4 | 287 | |||
FQ227764.1 | - | 3, 4 | 458 | |||
FQ230991.1 | - | 3, 4 | 287 | |||
FQ231108.1 | - | 3, 4 | 357 | |||
FQ231686.1 | - | 3, 4 | 299 | |||
FQ233099.1 | - | 3, 4 | 299 | |||
FQ234630.1 | - | 3, 4 | 299 | |||
L24896.1 | - | 3, 4 | 268 | |||
U37427.1 | - | 3, 4 | 385 | |||
X82679.1 | - | 3, 4 | 278 |