Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Npc2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_173118.2 | NP_775141.2 | 3 | 369 | ||
GenBank mRNA | AJ515237.1 | - | 3 | 304 | ||
BC058132.1 | - | 3 | 332 | 6920130 | ||
FQ214040.1 | - | 3 | 290 | |||
FQ216545.1 | - | 3 | 294 | |||
FQ219301.1 | - | 3 | 311 | |||
FQ219863.1 | - | 3 | 302 | |||
FQ220660.1 | - | 3 | 290 | |||
FQ220919.1 | - | 3 | 296 | |||
FQ228456.1 | - | 3 | 294 | |||
FQ229422.1 | - | 3 | 349 | |||
FQ229782.1 | - | 3 | 290 | |||
FQ229783.1 | - | 3 | 292 | |||
FQ230107.1 | - | 3 | 296 |