Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Fgf10 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_012951.1 | NP_037083.1 | 1 | 12 | ||
GenBank mRNA | D79215.1 | - | 1 | 12 |