Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Mobp | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_012720.1 | NP_036852.1 | 3 | 283 | ||
XM_006244065.2 | XP_006244127.1 | 3 | 367 | |||
XM_006244067.2 | XP_006244129.1 | 4 | 405 | |||
XM_006244069.2 | XP_006244131.1 | 4 | 405 | |||
XM_008766669.1 | XP_008764891.1 | 2 | 304 | |||
GenBank mRNA | AF157498.1 | - | 3 | 276 | ||
BC087694.1 | - | 2 | 237 | |||
BC161856.1 | - | 4 | 319 | 7320248 | ||
D28110.1 | - | 2 | 234 | |||
D28111.1 | - | 2 | 234 | |||
X87900.1 | - | 3 | 276 | |||
X89637.1 | - | 3 | 283 | |||
X89638.1 | - | 2 | 265 | |||
X90402.1 | - | 3 | 289 |