Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Antibodies
Cell Culture Media
Chemicals
Chromatography Columns and Cartridges
Lab Equipment
Lab Plasticware and Supplies
Microplates
Oligos, Primers, Probes and Genes
TaqMan Real-Time PCR Assays
Greener Products
See all product categories
Applications
Bioprocessing
Cell Culture and Transfection
Cell and Gene Therapy
Chromatography
Molecular Testing
Digital Solutions
DNA and RNA Extraction and Analysis
Spectroscopy, Elemental and Isotope Analysis
See all applications and techniques
Services
360° CDMO and CRO Solutions
CDMO Services
CRO Services
Custom Services
Enterprise Services
Financial and Leasing Services
Instrument Services
Lab Informatics
OEM and Commercial Supply
Training Services
Unity Lab Services
See all services
Help and Support
How to Order
Instrument Support
Learning Centers
Register for an Account
Technical Support Centers
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Contact Us
Quick Order
Order Status and Tracking
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Thermo Fisher Scientific
Search All
Search
Search button
Search button
Close
Order Status
Quick Order
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Aspire Member Program
Connect: Lab, Data, Apps
Custom Products & Projects
Services Central
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
See other TTR SIRNA Assays ›
Gene Symbol:
Ttr
Gene Name:
transthyretin
Gene Aliases:
Lr1, Tbpa
Chromosome Location:
Chr.18: 15532963 - 15542180 on Build Rnor_6.0
UniGene ID:
Rn.1404
Species:
Rat
siRNA ID
RSS351746
Size
20 nmol
20 nmol
Availability
Made To Order
Catalog #
1330001
Price
Your Price
Online offer:
Check your price ›
Add To Cart
Genomic Map
siRNA Details
More Information
Genomic Map
LOADING...
×
Back To Top
siRNA Details
Gene Symbol: Ttr
Interrogated Sequence
Translated Protein
Targeted Exon(s)
siRNA Location
IMAGE Clone ID
RefSeq
NM_012681.2
NP_036813.2
4
471
XM_006254457.1
XP_006254519.1
5
528
GenBank mRNA
AF479660.1
-
4
406
BC086946.1
-
4
364
7322736
FQ209711.1
-
4
386
FQ209724.1
-
4
389
FQ209731.1
-
4
389
FQ209737.1
-
4
389
FQ209743.1
-
4
389
FQ209798.1
-
4
389
FQ209827.1
-
4
388
FQ209853.1
-
4
389
FQ209868.1
-
4
389
FQ209884.1
-
4
389
FQ209896.1
-
4
382
FQ209911.1
-
4
389
FQ209919.1
-
4
388
FQ209931.1
-
4
386
FQ209949.1
-
4
389
FQ209960.1
-
4
388
FQ209974.1
-
4
389
FQ209986.1
-
4
389
FQ209992.1
-
4
386
FQ210005.1
-
4
389
FQ210012.1
-
4
388
FQ210053.1
-
4
389
FQ210063.1
-
4
389
FQ210074.1
-
4
384
FQ210079.1
-
4
389
FQ210088.1
-
4
389
FQ210092.1
-
4
389
FQ210104.1
-
4
388
FQ210128.1
-
4
389
FQ210130.1
-
4
389
FQ210144.1
-
4
389
FQ210148.1
-
4
389
FQ210149.1
-
4
389
FQ210166.1
-
4
389
FQ210168.1
-
4
389
FQ210188.1
-
4
389
FQ210192.1
-
4
384
FQ210193.1
-
4
389
FQ210202.1
-
4
389
FQ210212.1
-
4
389
FQ210220.1
-
4
389
FQ210221.1
-
4
389
FQ210257.1
-
4
389
FQ210271.1
-
4
389
FQ210283.1
-
4
386
FQ210289.1
-
4
384
FQ210300.1
-
4
389
FQ210301.1
-
4
389
FQ210303.1
-
4
389
FQ210314.1
-
4
389
FQ210319.1
-
4
389
FQ210324.1
-
4
387
FQ210363.1
-
4
389
FQ210365.1
-
4
388
FQ210370.1
-
4
389
FQ210380.1
-
4
389
FQ210383.1
-
4
386
FQ210386.1
-
839
FQ210389.1
-
4
389
FQ210423.1
-
4
388
FQ210429.1
-
4
389
FQ210431.1
-
4
389
FQ210434.1
-
4
389
FQ210451.1
-
4
389
FQ210464.1
-
4
384
FQ210465.1
-
4
389
FQ210502.1
-
4
389
FQ210512.1
-
4
388
FQ210545.1
-
4
389
FQ210547.1
-
4
389
FQ210548.1
-
4
386
FQ210552.1
-
4
384
FQ210579.1
-
4
388
FQ210580.1
-
4
386
FQ210584.1
-
4
389
FQ210588.1
-
4
389
FQ210614.1
-
4
389
FQ210642.1
-
4
389
FQ210659.1
-
4
388
FQ210663.1
-
4
386
FQ210667.1
-
4
389
FQ210680.1
-
4
384
FQ210689.1
-
4
389
FQ210691.1
-
4
389
FQ210711.1
-
4
386
FQ210744.1
-
4
389
FQ210747.1
-
4
387
FQ210750.1
-
4
388
FQ210759.1
-
4
389
FQ210777.1
-
4
386
FQ210778.1
-
4
389
FQ210779.1
-
4
389
FQ210782.1
-
4
389
FQ210786.1
-
4
389
FQ210844.1
-
4
389
FQ210853.1
-
4
386
FQ210869.1
-
4
389
FQ210887.1
-
4
389
FQ210913.1
-
4
389
FQ210915.1
-
4
389
FQ210971.1
-
4
389
FQ210974.1
-
4
389
FQ210975.1
-
4
389
FQ210993.1
-
4
389
FQ211000.1
-
4
389
FQ211011.1
-
4
389
FQ211023.1
-
4
389
FQ211028.1
-
4
386
FQ211048.1
-
4
389
FQ211066.1
-
4
389
FQ211087.1
-
4
389
FQ211100.1
-
4
389
FQ211107.1
-
4
389
FQ211134.1
-
4
388
FQ211143.1
-
4
389
FQ211145.1
-
4
389
FQ211149.1
-
4
389
FQ211173.1
-
4
389
FQ211184.1
-
4
395
FQ211191.1
-
4
389
FQ211204.1
-
4
389
FQ211212.1
-
4
389
FQ211228.1
-
4
389
FQ211233.1
-
4
391
FQ211237.1
-
4
386
FQ211240.1
-
4
389
FQ211260.1
-
4
389
FQ211262.1
-
4
387
FQ211291.1
-
4
386
FQ211302.1
-
4
389
FQ211335.1
-
4
389
FQ211341.1
-
4
389
FQ211348.1
-
4
386
FQ211403.1
-
4
389
FQ211448.1
-
4
389
FQ211620.1
-
4
389
FQ212160.1
-
4
389
FQ212302.1
-
4
389
FQ213410.1
-
4
474
FQ217460.1
-
4
389
FQ218087.1
-
4
389
FQ218127.1
-
4
390
FQ218128.1
-
4
385
FQ218131.1
-
4
389
FQ218133.1
-
4
389
FQ218134.1
-
4
389
FQ218142.1
-
4
384
FQ218150.1
-
4
389
FQ218178.1
-
4
389
FQ218187.1
-
4
389
FQ218296.1
-
4
389
FQ218301.1
-
4
389
FQ218306.1
-
4
389
FQ218313.1
-
4
389
FQ218333.1
-
4
384
FQ218513.1
-
4
384
FQ218589.1
-
4
389
FQ218636.1
-
4
389
FQ218738.1
-
4
389
FQ218740.1
-
4
389
FQ218747.1
-
4
389
FQ218749.1
-
4
389
FQ218758.1
-
4
314
FQ218786.1
-
4
389
FQ218849.1
-
4
389
FQ218866.1
-
4
389
FQ218996.1
-
4
389
FQ219153.1
-
4
386
FQ219387.1
-
4
389
FQ219428.1
-
4
389
FQ223520.1
-
4
384
FQ223553.1
-
4
389
FQ224434.1
-
4
389
K03252.1
-
4
369
X14876.1
-
4
362
Back To Top
More Information
Gene Ontology Categories:
Function(s)
Process(es)
thyroid hormone metabolic process
retinol metabolic process
thyroid hormone transport
hormone activity
hormone binding
identical protein binding
protein heterodimerization activity
thyroid hormone binding
Back To Top
Related Products
Anti-GAPDH, Mouse Monoclonal 6C5
Anti-β-Actin, Mouse Monoclonal AC-15
TaqMan
®
Gene Expression Cells-to-CT
™
Neon
™
Transfection System 100 µL Kit
Lipofectamine
™
RNAiMAX Transfection Reagent
Invivofectamine 2.0
Lipofectamine
™
2000 Transfection Reagent
Neon
™
Transfection System
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-666774c847-nqx77:80/100.66.134.75:80.
git-commit: 4f7bff5b1d64937db6e1726f71785770340a1908
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.34.0-2025.08.32-1.0