Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Antibodies
Cell Culture Media
Chemicals
Chromatography Columns and Cartridges
Lab Equipment
Lab Plasticware and Supplies
Microplates
Oligos, Primers, Probes and Genes
TaqMan Real-Time PCR Assays
Greener Products
See all product categories
Applications
Bioprocessing
Cell Culture and Transfection
Cell and Gene Therapy
Chromatography
Molecular Testing
Digital Solutions
DNA and RNA Extraction and Analysis
Spectroscopy, Elemental and Isotope Analysis
See all applications and techniques
Services
360° CDMO and CRO Solutions
CDMO Services
CRO Services
Custom Services
Enterprise Services
Financial and Leasing Services
Instrument Services
Lab Informatics
OEM and Commercial Supply
Training Services
Unity Lab Services
See all services
Help and Support
How to Order
Instrument Support
Learning Centers
Register for an Account
Technical Support Centers
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Contact Us
Quick Order
Order Status and Tracking
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Thermo Fisher Scientific
Search All
Search
Search button
Search button
Close
Order Status
Quick Order
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Aspire Member Program
Connect: Lab, Data, Apps
Custom Products & Projects
Services Central
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
See other ACTA1 SIRNA Assays ›
Gene Symbol:
Acta1
Gene Name:
actin, alpha 1, skeletal muscle
Gene Aliases:
-
Chromosome Location:
Chr.19: 56674072 - 56677084 on Build Rnor_6.0
UniGene ID:
Rn.82732
Species:
Rat
siRNA ID
RSS341453
Size
20 nmol
20 nmol
Availability
Made To Order
Catalog #
1330001
Price
Your Price
Online offer:
Check your price ›
Add To Cart
Genomic Map
siRNA Details
More Information
Genomic Map
LOADING...
×
Back To Top
siRNA Details
Gene Symbol: Acta1
Interrogated Sequence
Translated Protein
Targeted Exon(s)
siRNA Location
IMAGE Clone ID
RefSeq
NM_019212.2
NP_062085.1
5
855
GenBank mRNA
BC061974.1
-
5
855
5623909
FQ214598.1
-
5
865
FQ214610.1
-
3
409
FQ214623.1
-
5
863
FQ214659.1
-
5
866
FQ214676.1
-
5
865
FQ214734.1
-
5
865
FQ214743.1
-
5
864
FQ214761.1
-
5
863
FQ214785.1
-
5
863
FQ214786.1
-
5
867
FQ214822.1
-
5
865
FQ214841.1
-
5
864
FQ214842.1
-
5
865
FQ214857.1
-
5
868
FQ214858.1
-
5
864
FQ214860.1
-
5
866
FQ214866.1
-
5
866
FQ214883.1
-
5
863
FQ214904.1
-
5
864
FQ214921.1
-
5
865
FQ214948.1
-
5
864
FQ214949.1
-
5
865
FQ214969.1
-
5
864
FQ214975.1
-
5
866
FQ214976.1
-
5
864
FQ214987.1
-
5
864
FQ214990.1
-
5
864
FQ214993.1
-
5
864
FQ215012.1
-
5
865
FQ215025.1
-
5
866
FQ215038.1
-
5
863
FQ215044.1
-
5
864
FQ215058.1
-
5
865
FQ215089.1
-
5
865
FQ215136.1
-
5
864
FQ215153.1
-
5
864
FQ215185.1
-
5
864
FQ215204.1
-
5
862
FQ215212.1
-
5
866
FQ215241.1
-
5
865
FQ215249.1
-
5
865
FQ215256.1
-
5
864
FQ215277.1
-
5
865
FQ215282.1
-
5
864
FQ215298.1
-
5
865
FQ215324.1
-
5
864
FQ215366.1
-
5
865
FQ215380.1
-
5
866
FQ215393.1
-
5
864
FQ215396.1
-
5
864
FQ215398.1
-
5
864
FQ215402.1
-
5
864
FQ215403.1
-
5
864
FQ215424.1
-
5
865
FQ215431.1
-
5
865
FQ215464.1
-
5
864
FQ215465.1
-
5
864
FQ215470.1
-
5
864
FQ215488.1
-
5
864
FQ215518.1
-
5
873
FQ215554.1
-
5
864
FQ215618.1
-
5
865
FQ215628.1
-
5
866
FQ215631.1
-
3
478
FQ215655.1
-
5
862
FQ215659.1
-
5
863
FQ215673.1
-
5
864
FQ215674.1
-
5
864
FQ215690.1
-
5
865
FQ215692.1
-
5
865
FQ215785.1
-
5
864
FQ215790.1
-
5
866
FQ215809.1
-
5
864
FQ215813.1
-
5
864
FQ215836.1
-
5
864
FQ215839.1
-
5
864
FQ215841.1
-
5
865
FQ215851.1
-
5
864
FQ215852.1
-
5
862
FQ215864.1
-
5
865
FQ215866.1
-
5
865
FQ215888.1
-
5
863
FQ215893.1
-
5
866
FQ215896.1
-
5
864
FQ215924.1
-
3
556
FQ215964.1
-
5
864
FQ216035.1
-
5
864
FQ216043.1
-
5
864
FQ216050.1
-
5
864
FQ216224.1
-
5
865
FQ216246.1
-
5
865
FQ216276.1
-
5
864
FQ216287.1
-
5
864
FQ216291.1
-
5
865
FQ216293.1
-
5
865
FQ216305.1
-
5
865
FQ216329.1
-
5
864
FQ216350.1
-
5
864
FQ216352.1
-
5
864
FQ216360.1
-
5
865
FQ216403.1
-
5
864
FQ216430.1
-
5
865
FQ216509.1
-
5
862
FQ216603.1
-
5
866
FQ216606.1
-
5
864
FQ216607.1
-
5
860
FQ216620.1
-
5
865
FQ216639.1
-
5
864
FQ216644.1
-
5
865
FQ216675.1
-
5
864
FQ216676.1
-
5
864
FQ216703.1
-
5
864
FQ216721.1
-
5
864
FQ216725.1
-
5
860
FQ216735.1
-
1
104
FQ216739.1
-
5
864
FQ216767.1
-
5
864
FQ216776.1
-
5
865
FQ216777.1
-
5
864
FQ216793.1
-
5
864
FQ216799.1
-
5
862
FQ216801.1
-
5
864
FQ216819.1
-
5
862
FQ216918.1
-
3
507
FQ216999.1
-
1
63
FQ217061.1
-
5
866
FQ217091.1
-
5
864
FQ217173.1
-
5
864
FQ217188.1
-
5
864
FQ217194.1
-
5
864
FQ217280.1
-
5
984
FQ217329.1
-
5
864
FQ217353.1
-
5
864
FQ217441.1
-
5
864
FQ217443.1
-
5
866
FQ217475.1
-
5
865
FQ217525.1
-
5
864
FQ217535.1
-
5
864
FQ217572.1
-
5
864
FQ217615.1
-
5
864
FQ217629.1
-
5
865
FQ217641.1
-
5
861
FQ217668.1
-
1
113
FQ217762.1
-
5
864
FQ217893.1
-
5
865
FQ217919.1
-
5
864
FQ217937.1
-
5
864
FQ217990.1
-
5
864
FQ218005.1
-
2
264
FQ218019.1
-
5
865
FQ223563.1
-
5
865
FQ223681.1
-
5
868
FQ223686.1
-
5
864
FQ223702.1
-
5
865
FQ223727.1
-
3
562
FQ223778.1
-
3
378
FQ223898.1
-
5
865
FQ223926.1
-
5
864
FQ223929.1
-
5
863
FQ224082.1
-
5
864
FQ224691.1
-
5
862
FQ224840.1
-
5
862
FQ224842.1
-
5
864
FQ224862.1
-
5
864
FQ224886.1
-
5
863
FQ224890.1
-
5
862
FQ224936.1
-
5
865
FQ224946.1
-
3
392
FQ224971.1
-
5
864
Back To Top
More Information
Gene Ontology Categories:
Function(s)
Process(es)
response to mechanical stimulus
response to extracellular stimulus
response to lithium ion
cell growth
skeletal muscle thin filament assembly
skeletal muscle fiber adaptation
response to steroid hormone
skeletal muscle fiber development
ATP binding
Back To Top
Related Products
Anti-GAPDH, Mouse Monoclonal 6C5
Anti-β-Actin, Mouse Monoclonal AC-15
TaqMan
®
Gene Expression Cells-to-CT
™
Neon
™
Transfection System 100 µL Kit
Lipofectamine
™
RNAiMAX Transfection Reagent
Invivofectamine 2.0
Lipofectamine
™
2000 Transfection Reagent
Neon
™
Transfection System
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-666774c847-nqx77:80/100.66.134.75:80.
git-commit: 4f7bff5b1d64937db6e1726f71785770340a1908
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.34.0-2025.08.32-1.0