Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Kcnk1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_021688.3 | NP_067720.1 | 1 | 287 | ||
GenBank mRNA | AF022819.1 | - | 1 | 309 | ||
BC061807.1 | - | 1 | 287 | 5598415 |