Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Antibodies
Cell Culture Media
Chemicals
Chromatography Columns and Cartridges
Lab Equipment
Lab Plasticware and Supplies
Microplates
Oligos, Primers, Probes and Genes
TaqMan Real-Time PCR Assays
Greener Products
See all product categories
Applications
Bioprocessing
Cell Culture and Transfection
Cell and Gene Therapy
Chromatography
Molecular Testing
Digital Solutions
DNA and RNA Extraction and Analysis
Spectroscopy, Elemental and Isotope Analysis
See all applications and techniques
Services
360° CDMO and CRO Solutions
CDMO Services
CRO Services
Custom Services
Enterprise Services
Financial and Leasing Services
Instrument Services
Lab Informatics
OEM and Commercial Supply
Training Services
Unity Lab Services
See all services
Help and Support
How to Order
Instrument Support
Learning Centers
Register for an Account
Technical Support Centers
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Contact Us
Quick Order
Order Status and Tracking
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Thermo Fisher Scientific
Search All
Search
Search button
Search button
Close
Order Status
Quick Order
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Aspire Member Program
Connect: Lab, Data, Apps
Custom Products & Projects
Services Central
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
See other FZD1 SIRNA Assays ›
Gene Symbol:
Fzd1
Gene Name:
frizzled class receptor 1
Gene Aliases:
-
Chromosome Location:
Chr.4: 26470864 - 26475262 on Build Rnor_6.0
UniGene ID:
Rn.6575
Species:
Rat
siRNA ID
RSS339828
Size
20 nmol
20 nmol
Availability
Made To Order
Catalog #
1330001
Price
Your Price
Online offer:
Check your price ›
Add To Cart
Genomic Map
siRNA Details
More Information
Genomic Map
LOADING...
×
Back To Top
siRNA Details
Gene Symbol: Fzd1
Interrogated Sequence
Translated Protein
Targeted Exon(s)
siRNA Location
IMAGE Clone ID
RefSeq
NM_021266.3
NP_067089.2
1
1723
Back To Top
More Information
Set Membership:
G-protein coupled receptor
Gene Ontology Categories:
Function(s)
Process(es)
positive regulation of protein phosphorylation
membranous septum morphogenesis
muscular septum morphogenesis
outflow tract morphogenesis
G-protein coupled receptor signaling pathway
cell-cell signaling
Wnt signaling pathway
negative regulation of BMP signaling pathway
epithelial cell differentiation
negative regulation of catenin import into nucleus
autocrine signaling
non-canonical Wnt signaling pathway
response to drug
canonical Wnt signaling pathway involved in mesenchymal stem cell differentiation
canonical Wnt signaling pathway involved in osteoblast differentiation
regulation of osteoblast differentiation
negative regulation of transcription, DNA-templated
positive regulation of transcription, DNA-templated
lung alveolus development
positive regulation of sequence-specific DNA binding transcription factor activity
hard palate development
canonical Wnt signaling pathway
ventricular septum morphogenesis
cellular response to vitamin D
cellular response to growth factor stimulus
cellular response to peptide hormone stimulus
negative regulation of canonical Wnt signaling pathway
planar cell polarity pathway involved in neural tube closure
G-protein coupled receptor activity
receptor binding
frizzled binding
protein binding
Wnt-protein binding
PDZ domain binding
identical protein binding
protein homodimerization activity
Wnt-activated receptor activity
protein heterodimerization activity
Back To Top
Related Products
Anti-GAPDH, Mouse Monoclonal 6C5
Anti-β-Actin, Mouse Monoclonal AC-15
TaqMan
®
Gene Expression Cells-to-CT
™
Neon
™
Transfection System 100 µL Kit
Lipofectamine
™
RNAiMAX Transfection Reagent
Invivofectamine 2.0
Lipofectamine
™
2000 Transfection Reagent
Neon
™
Transfection System
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-666774c847-nqx77:80/100.66.134.75:80.
git-commit: 4f7bff5b1d64937db6e1726f71785770340a1908
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.34.0-2025.08.32-1.0