Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Gtpbp2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001013225.2 | NP_001013243.2 | 5, 6 | 819 | ||
XM_006244543.2 | XP_006244605.1 | 5, 6 | 861 | |||
XM_006244544.2 | XP_006244606.1 | 5, 6 | 651 | |||
XM_006244545.2 | XP_006244607.1 | 4, 5 | 2735 | |||
XM_006244546.2 | XP_006244608.1 | 5, 6 | 640 |