Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Txndc12 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001100840.1 | NP_001094310.1 | 4 | 371 | ||
GenBank mRNA | BC100255.1 | - | 4 | 313 | ||
BC158737.1 | - | 4 | 317 | |||
CV117277.1 | - | 4 | 608 | |||
FQ218354.1 | - | 4 | 268 |