Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Atp8a1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_006250974.2 | XP_006251036.1 | 29, 30 | 3048 | ||
XM_006250975.2 | XP_006251037.1 | 29, 30 | 3048 | |||
XM_006250976.2 | XP_006251038.1 | 28, 29 | 3033 | |||
XM_006250977.2 | XP_006251039.1 | 28, 29 | 3003 | |||
XM_006250980.2 | XP_006251042.1 | 25, 26 | 2534 | |||
XM_008770164.1 | XP_008768386.1 | 29, 30 | 3078 | |||
XM_008770165.1 | XP_008768387.1 | 26, 27 | 2786 |