Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Wdr7 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_023975.1 | NP_076465.1 | 14 | 2239 | ||
XM_008772171.1 | XP_008770393.1 | 15 | 2446 | |||
GenBank mRNA | AF192379.1 | - | 14 | 2239 | ||
AF305813.1 | - | 14 | 2239 |