Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Enterprise Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search Thermo Fisher Scientific

Search All
Search button Close
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          See other FASN SIRNA Assays ›
          Gene Symbol:
          Fasn
          Gene Name:
          fatty acid synthase
          Gene Aliases:
          -
          Chromosome Location:
          Chr.10: 109987735 - 110005901 on Build Rnor_6.0
          UniGene ID:
          Rn.9486
          Species:
          Rat
          siRNA ID RSS328904
          Size 20 nmol
          Availability Made To Order
          Catalog # 1330001
          Price 491.00
          Your Price
          Online offer:
          491.0
          Check your price ›
          • Genomic Map
          • siRNA Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          siRNA Details

          Gene Symbol: Fasn
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
          RefSeq NM_017332.1 NP_059028.1 34 5884
          GenBank mRNA M76767.1 - 34 5884
          X13415.1 - 32 5682
          X13527.1 - 1 45
          X62888.1 - 34 5889

          Back To Top

          More Information


          Gene Ontology Categories:

          Function(s) Process(es)

          osteoblast differentiation
          acetyl-CoA metabolic process
          fatty acid biosynthetic process
          lipid biosynthetic process
          oxidation-reduction process
          cellular response to interleukin-4
          fatty acid synthase activity
          [acyl-carrier-protein] S-acetyltransferase activity
          [acyl-carrier-protein] S-malonyltransferase activity
          3-oxoacyl-[acyl-carrier-protein] synthase activity
          3-oxoacyl-[acyl-carrier-protein] reductase (NADPH) activity
          3-hydroxypalmitoyl-[acyl-carrier-protein] dehydratase activity
          enoyl-[acyl-carrier-protein] reductase (NADPH, B-specific) activity
          oleoyl-[acyl-carrier-protein] hydrolase activity
          drug binding
          zinc ion binding
          myristoyl-[acyl-carrier-protein] hydrolase activity
          palmitoyl-[acyl-carrier-protein] hydrolase activity
          identical protein binding
          protein homodimerization activity
          poly(A) RNA binding
          enoyl-[acyl-carrier-protein] reductase (NADPH, A-specific) activity
          3-hydroxyoctanoyl-[acyl-carrier-protein] dehydratase activity
          NADPH binding

          Back To Top

          Related Products

          • Anti-GAPDH, Mouse Monoclonal 6C5
          • Anti-β-Actin, Mouse Monoclonal AC-15
          • TaqMan® Gene Expression Cells-to-CT™
          • Neon™ Transfection System 100 µL Kit
          • Lipofectamine™ RNAiMAX Transfection Reagent
          • Invivofectamine 2.0
          • Lipofectamine™ 2000 Transfection Reagent
          • Neon™ Transfection System
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us
          • Careers
          • Investors
          • News
          • Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2025 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-7967b4b575-m9zbg:80/100.66.134.75:80.
          git-commit: a47d3bef29014c2544d0032e3b149846d456d521
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.34.0-2025.08.32-1.0