Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Sypl2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001108563.1 | NP_001102033.1 | 3 | 401 | ||
GenBank mRNA | AB158471.1 | - | 3 | 219 |