Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ggnbp2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001004273.1 | NP_001004273.1 | 7 | 1251 | ||
XM_006247047.2 | XP_006247109.1 | 9 | 1465 | |||
XM_006247048.2 | XP_006247110.1 | 9 | 1458 | |||
XM_006247049.2 | XP_006247111.1 | 9 | 1462 | |||
XM_006247050.2 | XP_006247112.1 | 9 | 1455 | |||
XM_006247051.2 | XP_006247113.1 | 8 | 1337 | |||
XM_006247052.2 | XP_006247114.1 | 8 | 1331 | |||
XM_006247053.2 | XP_006247115.1 | 8 | 1334 | |||
XM_006247054.2 | XP_006247116.1 | 8 | 1327 | |||
XM_008768070.1 | XP_008766292.1 | 9 | 1439 | |||
XM_008768071.1 | XP_008766293.1 | 9 | 1409 | |||
GenBank mRNA | AY633742.1 | - | 7 | 1251 | ||
FQ220883.1 | - | 5 | 629 | |||
FQ227019.1 | - | 2 | 254 | |||
FQ232770.1 | - | 4 | 577 | |||
FQ234201.1 | - | 5 | 718 | |||
FQ234847.1 | - | 4 | 577 |