Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Fars2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001013139.1 | NP_001013157.1 | 5 | 1010 | ||
XM_006253823.2 | XP_006253885.1 | 5 | 1131 | |||
XM_006253825.2 | XP_006253887.1 | 5 | 1131 | |||
XM_006253826.2 | XP_006253888.1 | 5 | 1131 | |||
XM_008771572.1 | XP_008769794.1 | 6 | 1318 | |||
XM_008771573.1 | XP_008769795.1 | 6 | 1994 | |||
XM_008771574.1 | XP_008769796.1 | 5 | 1053 | |||
XM_008771575.1 | XP_008769797.1 | 6 | 1260 | |||
XM_008771576.1 | XP_008769798.1 | 6 | 1217 | |||
XM_008771577.1 | XP_008769799.1 | 5 | 1131 | |||
GenBank mRNA | BC078956.1 | - | 5 | 1010 | 7110464 | |
FQ232145.1 | - | 5 | 1050 |