Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Fars2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001013139.1 | NP_001013157.1 | 4 | 840 | ||
XM_006253823.2 | XP_006253885.1 | 4 | 961 | |||
XM_006253825.2 | XP_006253887.1 | 4 | 961 | |||
XM_006253826.2 | XP_006253888.1 | 4 | 961 | |||
XM_008771572.1 | XP_008769794.1 | 5 | 1148 | |||
XM_008771573.1 | XP_008769795.1 | 5 | 1824 | |||
XM_008771574.1 | XP_008769796.1 | 4 | 883 | |||
XM_008771575.1 | XP_008769797.1 | 5 | 1090 | |||
XM_008771576.1 | XP_008769798.1 | 5 | 1047 | |||
XM_008771577.1 | XP_008769799.1 | 4 | 961 | |||
GenBank mRNA | BC078956.1 | - | 4 | 840 | 7110464 | |
FQ232145.1 | - | 4 | 880 |