Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Med26 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001107300.2 | NP_001100770.2 | 3 | 1364 | ||
XM_006252800.2 | XP_006252862.1 | 3 | 6027 |