Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Osr1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001106716.2 | NP_001100186.1 | 1 | 166 | ||
XM_006239886.2 | XP_006239948.1 | 2 | 460 | |||
GenBank mRNA | BC159410.1 | - | 1 | 166 | 8366695 |