Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Wbp1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001106600.1 | NP_001100070.1 | 670 | |||
XM_006236712.2 | XP_006236774.1 | 729 | ||||
XM_006236713.2 | XP_006236775.1 | 638 | ||||
XM_008763021.1 | XP_008761243.1 | 531 | ||||
GenBank mRNA | BC079453.1 | - | 1227 | |||
BC098709.1 | - | 4 | 1224 | |||
BC158831.1 | - | 627 | 7379825 |
Gene Symbol: LOC103692173 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_008763090.1 | XP_008761312.1 | 5 | 710 | ||
XM_008763091.1 | XP_008761313.1 | 5 | 638 | |||
XM_008763092.1 | XP_008761314.1 | 4 | 629 | |||
XM_008763093.1 | XP_008761315.1 | 4 | 531 |
Gene Symbol: NEWGENE_1562258 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_008763087.1 | XP_008761309.1 | 8 | 2896 | ||
XM_008763089.1 | XP_008761311.1 | 8 | 3134 |