Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Enterprise Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search Thermo Fisher Scientific

Search All
Search button Close
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          See other APOH SIRNA Assays ›
          Gene Symbol:
          Apoh
          Gene Name:
          apolipoprotein H (beta-2-glycoprotein I)
          Gene Aliases:
          -
          Chromosome Location:
          Chr.10: 96640013 - 96653939 on Build Rnor_6.0
          UniGene ID:
          Rn.1824
          Species:
          Rat
          Species Specific ID (Flybase ID):
          -
          siRNA ID RSS305103
          Size 20 nmol
          Availability Made To Order
          Catalog # 1330001
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • siRNA Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          siRNA Details

          Gene Symbol: Apoh
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
          RefSeq NM_001009626.1 NP_001009626.1 3 326
          GenBank mRNA AM071386.1 - 3 285
          BC088180.1 - 3 326 7364928
          FQ209412.1 - 3 365
          FQ209491.1 - 3 352
          FQ209740.1 - 3 361
          FQ210182.1 - 3 352
          FQ210311.1 - 3 352
          FQ210756.1 - 3 368
          FQ210813.1 - 3 352
          FQ211328.1 - 3 325
          FQ218445.1 - 3 366
          FQ218454.1 - 3 352
          FQ218597.1 - 3 348
          FQ218599.1 - 3 352
          FQ218637.1 - 3 352
          FQ218785.1 - 3 368
          FQ218859.1 - 3 325
          FQ218865.1 - 3 352
          FQ218961.1 - 3 352
          FQ219138.1 - 3 417
          FQ219328.1 - 3 352
          FQ219380.1 - 3 353
          FQ219438.1 - 3 351
          FQ219481.1 - 3 365
          FQ219537.1 - 3 349
          FQ219580.1 - 3 366
          FQ219601.1 - 3 365
          FQ219674.1 - 3 352
          X15551.1 - 3 181

          Back To Top

          More Information


          Gene Ontology Categories:

          Function(s) Process(es)

          negative regulation of endothelial cell proliferation
          triglyceride metabolic process
          blood coagulation
          blood coagulation, intrinsic pathway
          negative regulation of endothelial cell migration
          negative regulation of angiogenesis
          regulation of blood coagulation
          negative regulation of blood coagulation
          organ regeneration
          plasminogen activation
          negative regulation of myeloid cell apoptotic process
          triglyceride transport
          negative regulation of smooth muscle cell apoptotic process
          positive regulation of lipoprotein lipase activity
          regulation of fibrinolysis
          negative regulation of fibrinolysis
          glycoprotein binding
          phospholipid binding
          heparin binding
          lipid binding
          identical protein binding
          lipoprotein lipase activator activity

          Back To Top

          Related Products

          • Anti-GAPDH, Mouse Monoclonal 6C5
          • Anti-β-Actin, Mouse Monoclonal AC-15
          • TaqMan® Gene Expression Cells-to-CT™
          • Neon™ Transfection System 100 µL Kit
          • Lipofectamine™ RNAiMAX Transfection Reagent
          • Invivofectamine 2.0
          • Lipofectamine™ 2000 Transfection Reagent
          • Neon™ Transfection System
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us
          • Careers
          • Investors
          • News
          • Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2025 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-956b84784-lf4xt:80/100.66.134.75:80.
          git-commit: 47437712f32dbf53ebfc670c484059c2aca19dcb
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.34.0-2025.08.32-1.0