Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Pabpc1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_134353.3 | NP_599180.1 | 8, 9 | 1450 | ||
GenBank mRNA | AJ298278.1 | - | 8, 9 | 1450 | ||
BC083176.1 | - | 8, 9 | 1694 | 7097075 | ||
FQ225136.1 | - | 1, 2 | 227 | |||
FQ225513.1 | - | 1, 2 | 163 | |||
FQ225570.1 | - | 2, 3 | 296 | |||
FQ225622.1 | - | 2, 3 | 346 | |||
FQ226477.1 | - | 3, 4 | 439 | |||
FQ226807.1 | - | 2, 3 | 366 | |||
FQ227116.1 | - | 1, 2 | 292 | |||
FQ227215.1 | - | 2, 3 | 388 | |||
FQ227400.1 | - | 3, 4 | 410 | |||
FQ227801.1 | - | 2, 3 | 337 | |||
FQ227809.1 | - | 1, 2 | 107 | |||
FQ228174.1 | - | 2, 3 | 351 | |||
FQ230227.1 | - | 3, 4 | 474 | |||
FQ230279.1 | - | 2, 3 | 325 | |||
FQ231168.1 | - | 5, 6 | 734 | |||
FQ231494.1 | - | 3, 4 | 469 | |||
FQ231712.1 | - | 1, 2 | 289 | |||
FQ231724.1 | - | 2, 3 | 362 | |||
FQ232521.1 | - | 2, 3 | 351 | |||
FQ232597.1 | - | 1, 2 | 227 | |||
FQ233029.1 | - | 2, 3 | 348 | |||
FQ233189.1 | - | 2, 3 | 363 | |||
FQ233212.1 | - | 1, 2 | 163 | |||
FQ233332.1 | - | 1, 2 | 161 | |||
FQ233392.1 | - | 1, 2 | 274 | |||
FQ233536.1 | - | 1, 2 | 163 | |||
FQ233735.1 | - | 1, 2 | 198 | |||
FQ233754.1 | - | 4, 5 | 579 | |||
FQ234055.1 | - | 1, 2 | 95 | |||
FQ234227.1 | - | 2, 3 | 349 | |||
FQ234338.1 | - | 1, 2 | 186 | |||
FQ234422.1 | - | 1, 2 | 274 | |||
FQ234731.1 | - | 5, 6 | 667 | |||
FQ235211.1 | - | 3, 4 | 393 |