Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Pabpc1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_134353.3 | NP_599180.1 | 6, 7 | 1069 | ||
GenBank mRNA | AJ298278.1 | - | 6, 7 | 1069 | ||
BC083176.1 | - | 6, 7 | 1313 | 7097075 | ||
FQ225555.1 | - | 3, 4 | 357 | |||
FQ226477.1 | - | 1, 2 | 58 | |||
FQ227400.1 | - | 1, 2 | 29 | |||
FQ230227.1 | - | 1, 2 | 93 | |||
FQ231126.1 | - | 3, 4 | 357 | |||
FQ231168.1 | - | 3, 4 | 354 | |||
FQ231494.1 | - | 1, 2 | 88 | |||
FQ233364.1 | - | 3, 4 | 357 | |||
FQ233754.1 | - | 2, 3 | 198 | |||
FQ234731.1 | - | 3, 4 | 286 | |||
FQ235211.1 | - | 1, 2 | 12 | |||
FQ235339.1 | - | 3, 4 | 346 |