Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Megf8 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_053628.2 | NP_446080.1 | 38 | 7027 | ||
XM_006228364.2 | XP_006228426.1 | 38 | 7373 | |||
GenBank mRNA | AB011534.1 | - | 6 | 941 |