Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: 1810014B01Rik | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK007490.1 | - | 2 | 114 | ||
AK012122.1 | - | 2 | 159 | |||
AK017634.1 | - | 169 | ||||
AK019015.1 | - | 2 | 116 | |||
AK049515.1 | - | 1 | 131 | |||
AK090016.1 | - | 2 | 156 |
Gene Symbol: 1190007I07Rik | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001135567.1 | NP_001129039.1 | 2 | 295 | ||
NM_001135568.1 | NP_001129040.1 | 2 | 166 | |||
NM_001135569.1 | NP_001129041.1 | 2 | 107 | |||
GenBank mRNA | AK004515.1 | - | 2 | 157 | ||
AK045413.1 | - | 2 | 179 | |||
BC038652.1 | - | 2 | 125 | |||
BC065417.1 | - | 2 | 129 | |||
DV649038.1 | - | 2 | 101 |