Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Phgdh | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_016966.3 | NP_058662.2 | 10 | 1304 | ||
GenBank mRNA | AK076815.1 | - | 10 | 1260 | ||
AK147946.1 | - | 10 | 1253 | |||
AK169684.1 | - | 10 | 1260 | |||
AW545294.1 | - | 3 | 564 | |||
BC086668.1 | - | 10 | 1212 | 30633243 | ||
BC110673.1 | - | 10 | 1246 | 6311873 | ||
L21027.1 | - | 9 | 1023 |
Gene Symbol: Gm6756 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | BB019743.2 | - | 1 | 33 |
Gene Symbol: Gm8096 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK015074.1 | - | 508 |