Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Pamr1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_173749.4 | NP_776110.3 | 2, 3 | 425 | ||
GenBank mRNA | AK031254.1 | - | 2, 3 | 298 | ||
AK088017.1 | - | 2, 3 | 324 | |||
BB620582.1 | - | 2, 3 | 292 | |||
BC031841.1 | - | 2, 3 | 379 | 3675412 | ||
BC057685.1 | - | 2, 3 | 379 | 3675412 | ||
BY266233.1 | - | 2, 3 | 376 |