Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Jun | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_010591.2 | NP_034721.1 | 1 | 1025 | ||
GenBank mRNA | AK132019.1 | - | 1 | 1000 | ||
AK133722.1 | - | 1 | 1005 | |||
AK133867.1 | - | 1 | 1007 | |||
AK135023.1 | - | 1 | 1004 | |||
AK140911.1 | - | 1 | 1000 | |||
AK159196.1 | - | 1 | 1000 | |||
AK159220.1 | - | 1 | 1005 | |||
BC002081.1 | - | 1 | 168 | 3493248 | ||
BC021888.1 | - | 1 | 959 | 3156091 | ||
BC032086.1 | - | 1 | 493 | |||
BC055887.1 | - | 1 | 1102 | |||
BC094032.1 | - | 2 | 887 | 4936392 | ||
J04115.1 | - | 1 | 990 | |||
X12740.1 | - | 1 | 688 | |||
X12761.1 | - | 1 | 428 |