Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Enterprise Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search Thermo Fisher Scientific

Search All
Search button Close
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          See other HSPA8 SIRNA Assays ›
          Gene Symbol:
          Hspa8 Eif4h
          Gene Name:
          heat shock protein 8
          eukaryotic translation initiation factor 4H
          Gene Aliases:
          2410008N15Rik, Hsc70, Hsc71, Hsc73, Hsp73, Hspa10
          AU018978, D5Ertd355e, E430026L18Rik, Ef4h, Wbscr1, Wscr1, mKIAA0038
          Chromosome Location:
          Chr.9: 40801273 - 40805199 on Build GRCm38
          Chromosome Location:
          Chr.5: 134619876 - 134639494 on Build GRCm38
          UniGene ID:
          Mm.290774
          Mm.27955
          Species:
          Mouse
          siRNA ID MSS274955
          Size 20 nmol
          Availability Made To Order
          Catalog # 1320001
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • siRNA Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          siRNA Details

          Gene Symbol: Hspa8
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
          RefSeq NM_031165.4 NP_112442.2 7 1543
          GenBank mRNA AK004608.1 - 4 2264
          AK035286.1 - 7 1541
          AK075935.1 - 7 1540
          AK081701.1 - 7 1542
          AK098074.1 - 7 1540
          AK135281.1 - 3 712
          AK145416.1 - 7 1463
          AK145579.1 - 7 1537
          AK146547.1 - 7 1547
          AK146708.1 - 7 1538
          AK146985.1 - 7 1541
          AK147864.1 - 7 1541
          AK149954.1 - 7 1538
          AK150474.1 - 7 1535
          AK150498.1 - 7 1535
          AK150701.1 - 7 1545
          AK150846.1 - 2 277
          AK150958.1 - 7 1538
          AK151041.1 - 7 1541
          AK151065.1 - 7 1541
          AK151127.1 - 7 1544
          AK151287.1 - 7 1541
          AK151435.1 - 7 1541
          AK151516.1 - 7 1547
          AK151537.1 - 7 1538
          AK151741.1 - 7 1541
          AK151775.1 - 7 1543
          AK151808.1 - 7 1544
          AK151865.1 - 7 1541
          AK151892.1 - 7 1543
          AK151948.1 - 7 1544
          AK151997.1 - 7 1541
          AK152315.1 - 7 1544
          AK152598.1 - 7 1541
          AK152697.1 - 7 1541
          AK152703.1 - 7 1541
          AK152768.1 - 7 1544
          AK152803.1 - 7 1547
          AK153032.1 - 7 1543
          AK153834.1 - 7 1541
          AK157831.1 - 7 1541
          AK159479.1 - 7 1541
          AK162775.1 - 7 1541
          AK163987.1 - 7 1541
          AK164000.1 - 7 1541
          AK166545.1 - 7 1543
          AK166643.1 - 7 1555
          AK166721.1 - 7 1538
          AK166767.1 - 7 1541
          AK166776.1 - 7 1544
          AK166808.1 - 7 1541
          AK166830.1 - 7 1539
          AK166846.1 - 7 1541
          AK166861.1 - 7 1544
          AK166873.1 - 7 1541
          AK166908.1 - 7 1544
          AK166910.1 - 7 1541
          AK166913.1 - 7 1541
          AK166933.1 - 7 1541
          AK167043.1 - 7 1543
          AK167121.1 - 7 1544
          AK167122.1 - 7 1541
          AK167134.1 - 7 1539
          AK167163.1 - 7 1543
          AK167218.1 - 7 1541
          AK167229.1 - 7 1542
          AK167254.1 - 7 1542
          AK167414.1 - 7 1541
          AK167845.1 - 7 1544
          AK167910.1 - 7 1544
          AK168435.1 - 7 1544
          AK168492.1 - 7 1543
          AK168511.1 - 5 1079
          AK168519.1 - 7 1541
          AK168542.1 - 7 1543
          AK168577.1 - 7 1542
          AK168625.1 - 7 1542
          AK168654.1 - 7 1542
          AK168711.1 - 7 1541
          AK168750.1 - 7 1544
          AK168776.1 - 7 1544
          AK168887.1 - 7 1543
          AK168934.1 - 7 1544
          AK169093.1 - 7 1541
          AK169179.1 - 7 1541
          AK169236.1 - 7 1541
          AK169293.1 - 7 1555
          AK169329.1 - 7 1544
          AK169594.1 - 7 1540
          AK177099.1 - 2 243
          AK178263.1 - 2 242
          AK178681.1 - 2 244
          AK186515.1 - 2 242
          AK197383.1 - 2 302
          AK197524.1 - 2 242
          AK199284.1 - 2 242
          AK204189.1 - 2 243
          AK204472.1 - 2 242
          AK215080.1 - 2 242
          AK217000.1 - 2 302
          AK217051.1 - 2 302
          BC006722.1 - 7 1565 3711399
          BC066191.1 - 7 1497 30467052
          BC085486.1 - 7 1529 30540914
          BC089322.1 - 7 1522 30538640
          BC089457.1 - 7 1500 30628505
          BC094900.1 - 7 1457 30063483
          BC106169.1 - 5 1221
          BC106193.1 - 7 1525 6329283
          M19141.1 - 7 1527
          U27129.1 - 7 1492
          Gene Symbol: Eif4h
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
          GenBank mRNA AK178505.1 - 236

          Back To Top

          More Information


          Gene Ontology Categories:

          Function(s) Process(es)

          transcription, DNA-templated
          regulation of transcription, DNA-templated
          mRNA processing
          protein folding
          RNA splicing
          protein refolding
          negative regulation of transcription, DNA-templated
          ATP metabolic process
          positive regulation of mRNA splicing, via spliceosome
          chaperone mediated protein folding requiring cofactor
          regulation of cell cycle
          chaperone-mediated protein folding
          clathrin coat disassembly
          negative regulation of fibril organization
          nucleotide binding
          G-protein coupled receptor binding
          receptor binding
          protein binding
          ATP binding
          ATPase activity
          enzyme binding
          MHC class II protein complex binding
          heat shock protein binding
          ubiquitin protein ligase binding
          ATPase activity, coupled
          ADP binding
          poly(A) RNA binding
          unfolded protein binding
          C3HC4-type RING finger domain binding

          Back To Top

          Related Products

          • Anti-GAPDH, Mouse Monoclonal 6C5
          • Anti-β-Actin, Mouse Monoclonal AC-15
          • TaqMan® Gene Expression Cells-to-CT™
          • Neon™ Transfection System 100 µL Kit
          • Lipofectamine™ RNAiMAX Transfection Reagent
          • Invivofectamine 2.0
          • Lipofectamine™ 2000 Transfection Reagent
          • Neon™ Transfection System
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us
          • Careers
          • Investors
          • News
          • Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2025 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-666774c847-nqx77:80/100.66.134.75:80.
          git-commit: 4f7bff5b1d64937db6e1726f71785770340a1908
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.34.0-2025.08.32-1.0