Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Plxnb2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001159521.2 | NP_001152993.1 | 14 | 2492 | ||
NM_001284506.1 | NP_001271435.1 | 13 | 2406 | |||
NM_138749.3 | NP_620088.2 | 14 | 2456 | |||
GenBank mRNA | AB093229.1 | - | 10 | 1040 | ||
AK147538.1 | - | 14 | 2457 | |||
AK154627.1 | - | 14 | 2407 | |||
BC007481.1 | - | 11 | 1254 | |||
BC036274.1 | - | 2 | 64 | |||
BC157960.1 | - | 14 | 2427 | 9088132 | ||
BC157961.1 | - | 14 | 2427 | 9088133 | ||
BC158086.1 | - | 14 | 2393 | 9088174 | ||
BC172162.1 | - | 14 | 2393 | 9054832 |