Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Eif4ebp3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_201256.4 | NP_957708.1 | 2, 3 | 406 | ||
GenBank mRNA | AY226182.1 | - | 2, 3 | 284 | ||
BC038258.1 | - | 2, 3 | 313 | |||
BC061242.1 | - | 2, 3 | 406 | 6743910 | ||
BC111107.1 | - | 2, 3 | 346 | 40055518 | ||
CX565744.1 | - | 1, 2 | 36 |
Gene Symbol: Ankhd1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK129284.1 | - | 5091 |
Gene Symbol: Maskbp3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK003441.1 | - | 7, 8 | 1472 | ||
BC016604.1 | - | 7, 8 | 1898 |