Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Tmem176a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001098271.1 | NP_001091741.1 | 5 | 552 | ||
NM_025326.4 | NP_079602.4 | 5 | 604 | |||
GenBank mRNA | AB063313.1 | - | 4 | 494 | ||
AK002546.1 | - | 5 | 580 | |||
AK079378.1 | - | 566 | ||||
AK150576.1 | - | 5 | 569 | |||
AK162991.1 | - | 3 | 1812 | |||
AK165660.1 | - | 5 | 578 | |||
AK180847.1 | - | 1 | 90 | |||
BC010831.1 | - | 5 | 548 | 4221626 | ||
CT010190.1 | - | 4 | 494 |