Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Mettl9 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_021554.2 | NP_067529.2 | 5 | 1270 | ||
GenBank mRNA | AB041664.1 | - | 5 | 1047 | ||
AJ278576.1 | - | 5 | 1270 | |||
AK003378.1 | - | 1 | 81 | |||
AK075586.1 | - | 5 | 1032 | |||
AK084366.1 | - | 996 | ||||
AK169951.1 | - | 4 | 686 | |||
AK206642.1 | - | 1 | 56 | |||
AK210143.1 | - | 1 | 56 | |||
AK211126.1 | - | 1 | 56 | |||
AK211493.1 | - | 1 | 56 | |||
AK215069.1 | - | 1 | 56 | |||
AK216352.1 | - | 1 | 56 | |||
AK216736.1 | - | 2 | 348 | |||
BC023188.1 | - | 4 | 711 | |||
BC068124.1 | - | 5 | 1064 | 5716972 | ||
BC082317.1 | - | 4 | 695 |