Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Pa2g4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_011119.3 | NP_035249.1 | 5 | 574 | ||
GenBank mRNA | AK002685.1 | - | 5 | 652 | ||
AK076286.1 | - | 5 | 579 | |||
AK144645.1 | - | 5 | 628 | |||
AK166936.1 | - | 5 | 583 | |||
AK168582.1 | - | 5 | 576 | |||
AK205907.1 | - | 1 | 33 | |||
AK207779.1 | - | 1 | 33 | |||
BC003777.1 | - | 5 | 564 | |||
BC005593.1 | - | 5 | 564 | |||
BC028926.1 | - | 5 | 557 | |||
BC037623.1 | - | 5 | 594 | |||
BC046532.1 | - | 5 | 528 | 5358060 | ||
BC086788.1 | - | 3 | 2780 | |||
U43918.1 | - | 5 | 733 | |||
X84789.1 | - | 5 | 639 |