Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ddx3x | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_010028.3 | NP_034158.1 | 11, 12 | 1262 | ||
GenBank mRNA | AK140990.1 | - | 11, 12 | 1263 | ||
AK163250.1 | - | 11, 12 | 1237 | |||
AK186622.1 | - | 1, 2 | 46 | |||
AK204341.1 | - | 1, 2 | 83 | |||
AK205639.1 | - | 1, 2 | 46 | |||
BC150862.1 | - | 11, 12 | 1216 | 9087773 | ||
BC172016.1 | - | 11, 12 | 1213 | 9054302 | ||
DQ858457.1 | - | 1, 2 | 51 | |||
DQ858458.1 | - | 1, 2 | 51 | |||
L25126.1 | - | 11, 12 | 1194 | |||
Z38117.1 | - | 11, 12 | 1246 |