Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ddx3x | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_010028.3 | NP_034158.1 | 2 | 155 | ||
GenBank mRNA | AK140990.1 | - | 2 | 156 | ||
AK163250.1 | - | 2 | 130 | |||
BB854842.1 | - | 2 | 157 | |||
BC150862.1 | - | 2 | 109 | 9087773 | ||
BC172016.1 | - | 2 | 109 | 9054302 | ||
L25126.1 | - | 2 | 87 | |||
Z38117.1 | - | 2 | 139 |