Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Gtf3c2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_027901.2 | NP_082177.2 | 14 | 2227 | ||
GenBank mRNA | AK004895.1 | - | 14 | 2005 | ||
AK012095.1 | - | 7 | 824 | |||
AK043944.1 | - | 14 | 1994 | |||
AK083585.1 | - | 14 | 2229 | |||
AK083636.1 | - | 13 | 1982 | |||
AK129033.1 | - | 13 | 1977 | |||
AK138760.1 | - | 1996 | ||||
AK149771.1 | - | 15 | 2124 | |||
AK151300.1 | - | 14 | 1992 | |||
AK152439.1 | - | 14 | 1992 | |||
AK159938.1 | - | 14 | 1995 | |||
BC026573.1 | - | 6 | 545 | |||
BC034369.1 | - | 14 | 2005 | 3994677 | ||
BC043100.1 | - | 14 | 2215 |