Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Gtf3c2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_027901.2 | NP_082177.2 | 7 | 1381 | ||
GenBank mRNA | AK004895.1 | - | 7 | 1159 | ||
AK035792.1 | - | 7 | 1147 | |||
AK043944.1 | - | 7 | 1148 | |||
AK083585.1 | - | 7 | 1383 | |||
AK083636.1 | - | 6 | 1136 | |||
AK129033.1 | - | 6 | 1131 | |||
AK138760.1 | - | 1150 | ||||
AK149771.1 | - | 8 | 1278 | |||
AK151300.1 | - | 7 | 1146 | |||
AK152439.1 | - | 7 | 1146 | |||
AK159938.1 | - | 7 | 1148 | |||
BC034369.1 | - | 7 | 1159 | 3994677 | ||
BC043100.1 | - | 7 | 1369 |