Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Pcyox1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_025823.4 | NP_080099.1 | 6 | 1165 | ||
GenBank mRNA | AK004799.2 | - | 6 | 1120 | ||
AK004840.1 | - | 6 | 1120 | |||
AK031585.1 | - | 6 | 1121 | |||
AK033373.1 | - | 6 | 1121 | |||
AK049412.1 | - | 6 | 1118 | |||
AK134528.1 | - | 6 | 1166 | |||
AK145366.1 | - | 6 | 1125 | |||
AK147190.1 | - | 6 | 1133 | |||
AK159981.1 | - | 6 | 1121 | |||
AK161616.1 | - | 6 | 1121 | |||
AK169209.1 | - | 6 | 1121 | |||
AK173058.1 | - | 6 | 1087 | |||
BC028308.1 | - | 6 | 1135 | 3486530 |