Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Pcyox1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_025823.4 | NP_080099.1 | 6 | 1527 | ||
GenBank mRNA | AK004799.2 | - | 6 | 1482 | ||
AK004840.1 | - | 6 | 1482 | |||
AK031585.1 | - | 6 | 1483 | |||
AK033373.1 | - | 6 | 1483 | |||
AK049412.1 | - | 6 | 1480 | |||
AK134528.1 | - | 6 | 1528 | |||
AK145366.1 | - | 6 | 1487 | |||
AK147190.1 | - | 6 | 1495 | |||
AK159981.1 | - | 6 | 1483 | |||
AK161616.1 | - | 6 | 1483 | |||
AK169209.1 | - | 6 | 1483 | |||
AK173058.1 | - | 6 | 1449 | |||
AK196086.1 | - | 1 | 12 | |||
BC006056.1 | - | 4 | 1138 | |||
BC028308.1 | - | 6 | 1497 | 3486530 |