Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Phf21a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001109690.1 | NP_001103160.1 | 3, 4 | 141 | ||
NM_001109691.1 | NP_001103161.1 | 7, 8 | 549 | |||
NM_138755.2 | NP_620094.2 | 6, 7 | 555 | |||
GenBank mRNA | AB105178.1 | - | 6, 7 | 461 | ||
AK037174.1 | - | 7, 8 | 550 | |||
AK129424.1 | - | 5, 6 | 1559 | |||
AK137085.1 | - | 2, 3 | 102 | |||
BC019181.1 | - | 6, 7 | 410 | 5008954 | ||
BY303333.1 | - | 5, 6 | 325 | |||
CX217243.1 | - | 6, 7 | 555 |