Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Smad6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_008542.3 | NP_032568.3 | 2, 3 | 1915 | ||
GenBank mRNA | AB221689.1 | - | 2, 3 | 880 | ||
AF010133.1 | - | 2, 3 | 880 | |||
AK004671.1 | - | 2, 3 | 361 | |||
AK046602.1 | - | 2, 3 | 1918 | |||
BC047280.1 | - | 2, 3 | 1177 | 6477199 |