Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Usp33 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001076676.2 | NP_001070144.1 | 19, 20 | 2400 | ||
NM_001252486.1 | NP_001239415.1 | 19, 20 | 2400 | |||
NM_133247.3 | NP_573510.2 | 19, 20 | 2424 | |||
GenBank mRNA | AF383174.1 | - | 19, 20 | 2413 | ||
AK082251.1 | - | 19, 20 | 2383 | |||
AK082943.1 | - | 19, 20 | 2383 | |||
AK087467.1 | - | 19, 20 | 2401 | |||
AK122442.1 | - | 16, 17 | 4882 | |||
AK143844.1 | - | 19, 20 | 2392 | |||
BC005506.1 | - | 19, 20 | 2353 | |||
BC031366.1 | - | 11, 12 | 1540 | |||
BC036980.1 | - | 12, 13 | 1600 | |||
BC049870.1 | - | 6, 7 | 665 | |||
BC067399.1 | - | 18, 19 | 2266 | |||
BC089315.1 | - | 19, 20 | 2346 |